Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006786.hg.1 |
TC02000443.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006786.hg.1 |
ENST00000264448.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006786.hg.1 |
ENST00000409009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006786.hg.1 |
NM_015120.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155618.hg.1 |
ENST00000423048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155618.hg.1 |
ENST00000484298.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155618.hg.1 |
ENST00000613296.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155618.hg.1 |
ENST00000614410.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155618.hg.1 |
ENST00000620466.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155618.hg.1 |
NM_015120.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155618.hg.1 |
ENST00000423048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155618.hg.1 |
ENST00000484298.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155618.hg.1 |
ENST00000613296.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155618.hg.1 |
ENST00000614410.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155618.hg.1 |
ENST00000620466.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155618.hg.1 |
NM_015120.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155618.hg.1 |
TC0200008047.hg.1 |
Clariom D TC |