Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019231.hg.1 |
ENST00000254260.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019231.hg.1 |
ENST00000400226.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019231.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019231.hg.1 |
TC19001394.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019233.hg.1 |
ENST00000254260.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019233.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019233.hg.1 |
TC19001394.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019232.hg.1 |
ENST00000254260.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019232.hg.1 |
ENST00000400226.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019232.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019232.hg.1 |
TC19001394.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000585641.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000585641.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000585641.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000588683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000588683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000588683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000588683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000588683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000591502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000591502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000591502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000591502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000592247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000592247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000592247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
ENST00000592247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900185055.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185055.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185055.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900185055.hg.1 |
TC1900010375.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900185055.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185055.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185055.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900185054.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185054.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185054.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900185054.hg.1 |
TC1900010375.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900185054.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185054.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185054.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900185056.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900185056.hg.1 |
TC1900010375.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900185056.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900185053.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185053.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185053.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900185053.hg.1 |
TC1900010375.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900185053.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185053.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185053.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000254260.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000544458.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000585641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000585641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000585641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000588388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000588683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000588683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000588683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000588683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000588683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000591502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000591502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000591502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000591502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000592247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000592247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000592247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
ENST00000592247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010375.hg.1 |
NM_033103.4 |
Refseq |