Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000233596.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000233596.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000233596.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000233596.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000233596.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000233596.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000233596.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000233596.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000233596.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000233596.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000233596.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000233596.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000233596.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000395479.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000395479.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000395479.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000395479.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000395479.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000395479.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000395479.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000395484.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000395484.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000395484.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000395484.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000395484.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000395484.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000591735.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000591735.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000591735.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000591735.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000591735.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
ENST00000591735.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
NM_138393.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
NM_138393.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
NM_138393.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
NM_138393.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
NM_138393.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
NM_138393.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
NM_138393.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
NM_138393.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
NM_138393.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
NM_138393.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
NM_138393.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006537.hg.1 |
NM_138393.3 |
Refseq |