Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
NR_001275_at |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8158988 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8158989 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8158990 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8158987 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
207412_PM_x_at |
- |
- |
Affymetrix Human Genome U219 Array |
11760015_at |
- |
- |
Affymetrix Human Genome U219 Array |
11760015_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012186.hg.1 |
ENST00000411440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012186.hg.1 |
ENST00000411440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012186.hg.1 |
ENST00000411440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012186.hg.1 |
ENST00000411440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012186.hg.1 |
ENST00000411440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012186.hg.1 |
ENST00000411440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012186.hg.1 |
ENST00000411440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012186.hg.1 |
ENST00000411440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012186.hg.1 |
ENST00000421286.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012186.hg.1 |
ENST00000421286.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012186.hg.1 |
ENST00000421286.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012186.hg.1 |
ENST00000421286.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012186.hg.1 |
ENST00000421286.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012186.hg.1 |
ENST00000421286.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012186.hg.1 |
ENST00000421286.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012186.hg.1 |
ENST00000624767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012186.hg.1 |
ENST00000624767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012186.hg.1 |
ENST00000624767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012186.hg.1 |
NR_001275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012186.hg.1 |
NR_001275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012186.hg.1 |
NR_001275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012186.hg.1 |
NR_001275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012186.hg.1 |
NR_001275.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012186.hg.1 |
NR_001275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012186.hg.1 |
ENST00000411440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012186.hg.1 |
ENST00000411440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012186.hg.1 |
ENST00000411440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012186.hg.1 |
ENST00000411440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012186.hg.1 |
ENST00000411440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012186.hg.1 |
ENST00000411440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012186.hg.1 |
ENST00000411440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012186.hg.1 |
ENST00000411440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012186.hg.1 |
ENST00000421286.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012186.hg.1 |
ENST00000421286.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012186.hg.1 |
ENST00000421286.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012186.hg.1 |
ENST00000421286.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012186.hg.1 |
ENST00000421286.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012186.hg.1 |
ENST00000421286.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012186.hg.1 |
ENST00000421286.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012186.hg.1 |
ENST00000624767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012186.hg.1 |
ENST00000624767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012186.hg.1 |
ENST00000624767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012186.hg.1 |
NR_001275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012186.hg.1 |
NR_001275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012186.hg.1 |
NR_001275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012186.hg.1 |
NR_001275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012186.hg.1 |
NR_001275.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012186.hg.1 |
NR_001275.2 |
Refseq |
Human Genome U133 Plus 2.0 Array |
207412_x_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
207412_PM_x_at |
- |
- |
Human Genome U133A 2.0 Array |
207412_x_at |
- |
- |
Human Genome U133A 2.0 Array |
207412_PM_x_at |
- |
- |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) |
NR_001275_at |
- |
- |
AffymetrixPathwork Diagnostics Pathchip |
pw577362 |
- |
- |
Human Genome U95 Set |
31483_PM_g_at |
- |
- |
Human Genome U95 Set |
31483_g_at |
- |
- |
Human Genome U95 Set |
31482_PM_at |
- |
- |
Human Genome U95 Set |
31482_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
207412_x_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
207412_x_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
207412_x_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
207412_x_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
207412_x_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
207412_x_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
207412_x_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
207412_x_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
207412_x_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
207412_x_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
207412_x_at3 |
- |
- |
Human Genome U133 Set |
207412_x_at |
- |
- |
Human Genome U133 Set |
207412_PM_x_at |
- |
- |