Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013543.hg.1 |
TC09001034.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013543.hg.1 |
ENST00000297623.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013543.hg.1 |
ENST00000379124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013543.hg.1 |
ENST00000379126.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013543.hg.1 |
ENST00000379127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013543.hg.1 |
ENST00000379133.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013543.hg.1 |
ENST00000481295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013543.hg.1 |
NM_001252195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013543.hg.1 |
NM_032596.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013543.hg.1 |
NM_147168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013543.hg.1 |
NM_147169.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000297623.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000297623.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000297623.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000297623.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000297623.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000297623.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000297623.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000297623.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000297623.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000297623.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000297623.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379126.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379133.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379133.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379133.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379133.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379133.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379133.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379133.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000379133.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000444429.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000444429.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000444429.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000444429.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000481295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000481295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000481295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
ENST00000481295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_001252195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_001252195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_001252195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_001252195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_001252195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_001252195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_001252195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_032596.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_032596.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_032596.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_032596.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_032596.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_032596.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_032596.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_032596.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_032596.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_032596.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_032596.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_147168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_147168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_147168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_147168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_147168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_147168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_147168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_147168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_147169.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_147169.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_147169.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_147169.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_147169.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_147169.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009895.hg.1 |
NM_147169.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167808.hg.1 |
ENST00000297623.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167808.hg.1 |
ENST00000379124.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167808.hg.1 |
ENST00000379126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167808.hg.1 |
ENST00000379127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167808.hg.1 |
ENST00000379133.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167808.hg.1 |
ENST00000481295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167808.hg.1 |
NM_001252195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167808.hg.1 |
NM_032596.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167808.hg.1 |
NM_147168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167808.hg.1 |
NM_147169.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167808.hg.1 |
ENST00000297623.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167808.hg.1 |
ENST00000379124.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167808.hg.1 |
ENST00000379126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167808.hg.1 |
ENST00000379127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167808.hg.1 |
ENST00000379133.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167808.hg.1 |
ENST00000481295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167808.hg.1 |
NM_001252195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167808.hg.1 |
NM_032596.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167808.hg.1 |
NM_147168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167808.hg.1 |
NM_147169.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167808.hg.1 |
TC0900009895.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000297623.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000297623.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000297623.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000297623.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000297623.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000297623.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000297623.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000297623.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000297623.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000297623.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000297623.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379124.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379124.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379124.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379124.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379124.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379124.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379124.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379126.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379133.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379133.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379133.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379133.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379133.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379133.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379133.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000379133.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000444429.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000444429.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000444429.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000444429.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000481295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000481295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000481295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
ENST00000481295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_001252195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_001252195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_001252195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_001252195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_001252195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_001252195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_001252195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_032596.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_032596.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_032596.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_032596.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_032596.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_032596.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_032596.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_032596.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_032596.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_032596.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_032596.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_147168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_147168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_147168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_147168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_147168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_147168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_147168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_147168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_147169.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_147169.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_147169.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_147169.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_147169.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_147169.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009895.hg.1 |
NM_147169.2 |
Refseq |