Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013921.hg.1 |
TC04001007.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013921.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013921.hg.1 |
ENST00000429301.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013921.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013570.hg.1 |
ENST00000411606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013570.hg.1 |
ENST00000411606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013570.hg.1 |
ENST00000411606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013570.hg.1 |
ENST00000411606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013570.hg.1 |
ENST00000411606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013570.hg.1 |
ENST00000411606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013570.hg.1 |
ENST00000411606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013570.hg.1 |
ENST00000411606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013570.hg.1 |
ENST00000442005.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013918.hg.1 |
TC04001007.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013918.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013918.hg.1 |
ENST00000429301.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013918.hg.1 |
ENST00000514056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013918.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013917.hg.1 |
TC04001007.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013917.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013917.hg.1 |
ENST00000514056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013917.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000504920.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000504920.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000504920.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000504920.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000504920.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000504920.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000504920.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000504920.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000504920.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000509696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000509696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000509696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000509696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000509696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000509696.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000514056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000514056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000514056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
ENST00000514056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009936.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013915.hg.1 |
TC04001007.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013915.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013915.hg.1 |
ENST00000429301.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013915.hg.1 |
ENST00000514056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013915.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400168285.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168285.hg.1 |
ENST00000504920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168285.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400168285.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168285.hg.1 |
ENST00000504920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168285.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400168285.hg.1 |
TC0400009936.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700013570.hg.1 |
ENST00000411606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013570.hg.1 |
ENST00000411606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013570.hg.1 |
ENST00000411606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013570.hg.1 |
ENST00000411606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013570.hg.1 |
ENST00000411606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013570.hg.1 |
ENST00000411606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013570.hg.1 |
ENST00000411606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013570.hg.1 |
ENST00000411606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013570.hg.1 |
ENST00000442005.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168284.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168284.hg.1 |
ENST00000504920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168284.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400168284.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168284.hg.1 |
ENST00000504920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168284.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400168284.hg.1 |
TC0400009936.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000504920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000504920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000504920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000504920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000504920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000504920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000504920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000504920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000504920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000509696.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000509696.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000509696.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000509696.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000509696.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000509696.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000514056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000514056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000514056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
ENST00000514056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009936.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400168281.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168281.hg.1 |
ENST00000504920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168281.hg.1 |
ENST00000509696.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168281.hg.1 |
ENST00000514056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168281.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400168281.hg.1 |
ENST00000264954.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168281.hg.1 |
ENST00000504920.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168281.hg.1 |
ENST00000509696.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168281.hg.1 |
ENST00000514056.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168281.hg.1 |
NM_025196.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400168281.hg.1 |
TC0400009936.hg.1 |
Clariom D TC |