Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010472.hg.1 |
ENST00000632950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010472.hg.1 |
ENST00000632950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010563.hg.1 |
ENST00000450948.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010563.hg.1 |
ENST00000583555.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010563.hg.1 |
NR_049827.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010806.hg.1 |
ENST00000390626.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010806.hg.1 |
ENST00000390626.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010806.hg.1 |
ENST00000390626.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010563.hg.1 |
ENST00000450948.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010563.hg.1 |
ENST00000583555.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010563.hg.1 |
NR_049827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400177785.hg.1 |
ENST00000583555.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400177785.hg.1 |
NR_049827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400177785.hg.1 |
ENST00000583555.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400177785.hg.1 |
NR_049827.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400177785.hg.1 |
TC1400010563.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400177786.hg.1 |
TC1400010563.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400177788.hg.1 |
TC1400010563.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400010472.hg.1 |
ENST00000632950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010472.hg.1 |
ENST00000632950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400177787.hg.1 |
ENST00000450948.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400177787.hg.1 |
ENST00000450948.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400177787.hg.1 |
TC1400010563.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400010806.hg.1 |
ENST00000390626.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010806.hg.1 |
ENST00000390626.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010806.hg.1 |
ENST00000390626.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400177784.hg.1 |
TC1400010563.hg.1 |
Clariom D TC |