Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020744.3 | NP_065795.1 | 11 - 12 | 1256 | 105 | |
XM_005264456.2 | XP_005264513.1 | 11 - 12 | 1058 | 105 | ||
XM_005264459.2 | XP_005264516.1 | 12 - 13 | 1529 | 105 | ||
GenBank mRNA | AK027304.1 | - | 11 - 12 | 1054 | 105 | |
BC004227.2 | - | 11 - 12 | 1084 | 3542706 | 105 | |
BC053631.1 | - | 11 - 12 | 1242 | 6472900 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |