Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12028048.hg.1 |
ENST00000261655.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12028048.hg.1 |
ENST00000392375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12028048.hg.1 |
ENST00000535703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12028048.hg.1 |
ENST00000536002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12028048.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12028048.hg.1 |
TC12002130.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000536632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000536632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000536632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000536632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000536632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000536632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000540658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000540658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000540658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000540658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000540658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000544568.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000544568.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000544568.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000544568.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
ENST00000623766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204899.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204899.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204899.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204899.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204899.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204899.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204899.hg.1 |
TC1200012867.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000261655.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000535703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000536632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000536632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000536632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000536632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000536632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000536632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000540658.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000540658.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000540658.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000540658.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000540658.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000544568.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000544568.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000544568.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000544568.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
ENST00000623766.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012867.hg.1 |
NM_015347.5 |
Refseq |