Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000404147.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000404147.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000404147.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000404147.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000404147.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000404147.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000404147.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000404147.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000404147.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000404147.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000462902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000462902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000462902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000462902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000462902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000462902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000479032.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000479032.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000479032.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000479032.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000479032.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000479032.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000479032.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000479032.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000479032.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000479032.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000524733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000524733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000524733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000524733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000524733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000524733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000527430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000527430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000527430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000527430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000527430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000527430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000531401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000531401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000531401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000531401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000531401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000531401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000531401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000531401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000533015.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000533015.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000533015.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000533015.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000533015.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
ENST00000533015.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013184.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023068.hg.1 |
ENST00000275517.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023068.hg.1 |
ENST00000404147.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023068.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023068.hg.1 |
TC11001924.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100204545.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204545.hg.1 |
ENST00000404147.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204545.hg.1 |
ENST00000462902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204545.hg.1 |
ENST00000479032.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204545.hg.1 |
ENST00000524733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204545.hg.1 |
ENST00000527430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204545.hg.1 |
ENST00000531401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204545.hg.1 |
ENST00000533015.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204545.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204545.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204545.hg.1 |
ENST00000404147.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204545.hg.1 |
ENST00000462902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204545.hg.1 |
ENST00000479032.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204545.hg.1 |
ENST00000524733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204545.hg.1 |
ENST00000527430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204545.hg.1 |
ENST00000531401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204545.hg.1 |
ENST00000533015.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204545.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204545.hg.1 |
TC1100013184.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000275517.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000404147.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000404147.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000404147.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000404147.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000404147.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000404147.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000404147.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000404147.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000404147.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000404147.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000462902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000462902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000462902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000462902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000462902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000462902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000479032.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000479032.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000479032.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000479032.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000479032.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000479032.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000479032.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000479032.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000479032.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000479032.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000524733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000524733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000524733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000524733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000524733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000524733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000527430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000527430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000527430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000527430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000527430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000527430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000531401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000531401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000531401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000531401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000531401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000531401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000531401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000531401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000533015.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000533015.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000533015.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000533015.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000533015.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
ENST00000533015.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
NM_080668.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013184.hg.1 |
NM_080668.3 |
Refseq |