Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007627.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007627.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007627.hg.1 |
TC10000672.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000457045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000457045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000485048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000485048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000485048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000485048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000485048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000485048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000457045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000457045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000485048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000485048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000485048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000485048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000485048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000485048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157877.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157877.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157877.hg.1 |
ENST00000225235.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157877.hg.1 |
NM_015188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157877.hg.1 |
TC1000008502.hg.1 |
Clariom D TC |