Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022221.hg.1 |
TC04001592.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022221.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022221.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022223.hg.1 |
TC04001592.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022223.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000502786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000502786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000502786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000514787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000514787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
ENST00000514787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022222.hg.1 |
TC04001592.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022222.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04022222.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000502786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000502786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000502786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000514787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000514787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
ENST00000514787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011978.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400182190.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400182190.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400182190.hg.1 |
TC0400011978.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400182187.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182187.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400182187.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182187.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400182187.hg.1 |
TC0400011978.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400182188.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182188.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400182188.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182188.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400182188.hg.1 |
TC0400011978.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400182189.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182189.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400182189.hg.1 |
ENST00000442267.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400182189.hg.1 |
NM_015130.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400182189.hg.1 |
TC0400011978.hg.1 |
Clariom D TC |