Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTSL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301110.1 | NP_001288039.1 | 23 - 24 | 4198 | 75 | |
NM_207517.2 | NP_997400.2 | 23 - 24 | 4198 | 75 | ||
XM_011521822.2 | XP_011520124.1 | 24 - 25 | 4270 | 75 | ||
XM_011521823.2 | XP_011520125.1 | 24 - 25 | 4270 | 75 | ||
XM_011521824.2 | XP_011520126.1 | 24 - 25 | 4270 | 75 | ||
XM_011521825.2 | XP_011520127.1 | 24 - 25 | 4270 | 75 | ||
XM_017022434.1 | XP_016877923.1 | 24 - 25 | 4270 | 75 | ||
XM_017022435.1 | XP_016877924.1 | 23 - 24 | 4066 | 75 | ||
GenBank mRNA | AB033059.1 | - | 7 - 8 | 1972 | 75 | |
AK308845.1 | - | 23 - 24 | 4189 | 75 | ||
BC128389.1 | - | 23 - 24 | 4196 | 40125494 | 75 | |
BC128390.1 | - | 23 - 24 | 4196 | 40125501 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons |