Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018767.hg.1 |
TC05001329.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018767.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018767.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018766.hg.1 |
TC05001329.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018766.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018766.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010662.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500174511.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174511.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500174511.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174511.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500174511.hg.1 |
TC0500010662.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500174512.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174512.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500174512.hg.1 |
ENST00000303230.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174512.hg.1 |
NM_021072.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500174512.hg.1 |
TC0500010662.hg.1 |
Clariom D TC |