Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031687.hg.1 |
TC03001744.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031687.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03031687.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000518740.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000518740.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000518740.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300208715.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300208715.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300208715.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300208715.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300208715.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300208715.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300208715.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300208715.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300208715.hg.1 |
TC0300014039.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000518740.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000518740.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000518740.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000523403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
ENST00000524230.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_001173513.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014039.hg.1 |
NM_052883.1 |
Refseq |