Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
NM_153478_at |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
NM_001102576_at |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8170589 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8170578 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8175713 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8175720 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8170579 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8170582 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8170588 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8175711 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8170577 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8170576 |
- |
- |
Affymetrix Human Genome U219 Array |
11730715_x_at |
- |
- |
Affymetrix Human Genome U219 Array |
11730715_PM_x_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009585.hg.1 |
TC0X000741.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009585.hg.1 |
ENST00000370287.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009585.hg.1 |
ENST00000370291.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009585.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009585.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009585.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009589.hg.1 |
TC0X000741.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009589.hg.1 |
ENST00000370287.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009589.hg.1 |
ENST00000370291.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009589.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009589.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009589.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
ENST00000361211.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
ENST00000361211.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
ENST00000361211.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
ENST00000361211.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
ENST00000370287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
ENST00000370287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
ENST00000370287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
ENST00000370287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
ENST00000370287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
ENST00000370287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
ENST00000370291.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
ENST00000370291.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
ENST00000370291.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
ENST00000370291.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
ENST00000370291.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
ENST00000370291.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011134.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
ENST00000370287.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
ENST00000370287.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
ENST00000370287.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
ENST00000370287.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
ENST00000370291.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
ENST00000370287.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
ENST00000370291.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
ENST00000370287.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
ENST00000370291.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
ENST00000370291.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
ENST00000370287.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X000741.hg.1 |
ENST00000370291.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009577.hg.1 |
TC0X000741.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009577.hg.1 |
ENST00000370287.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009577.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009577.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009583.hg.1 |
TC0X000741.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009583.hg.1 |
ENST00000370287.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009583.hg.1 |
ENST00000370291.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009583.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009581.hg.1 |
TC0X000741.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009581.hg.1 |
ENST00000370287.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009581.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009581.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009581.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009582.hg.1 |
TC0X000741.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009582.hg.1 |
ENST00000370287.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009582.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009582.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009582.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009587.hg.1 |
TC0X000741.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009587.hg.1 |
ENST00000370287.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009587.hg.1 |
ENST00000370291.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009587.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009587.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009587.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175457.hg.1 |
ENST00000370287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175457.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175457.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175457.hg.1 |
ENST00000370287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175457.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175457.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175457.hg.1 |
TC0X00011134.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00175446.hg.1 |
ENST00000361211.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175446.hg.1 |
ENST00000370287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175446.hg.1 |
ENST00000370291.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175446.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175446.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175446.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175446.hg.1 |
ENST00000361211.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175446.hg.1 |
ENST00000370287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175446.hg.1 |
ENST00000370291.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175446.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175446.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175446.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175446.hg.1 |
TC0X00011134.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
ENST00000361211.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
ENST00000361211.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
ENST00000361211.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
ENST00000361211.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
ENST00000370287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
ENST00000370287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
ENST00000370287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
ENST00000370287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
ENST00000370287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
ENST00000370287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
ENST00000370291.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
ENST00000370291.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
ENST00000370291.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
ENST00000370291.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
ENST00000370291.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
ENST00000370291.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011134.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175448.hg.1 |
ENST00000361211.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175448.hg.1 |
ENST00000370287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175448.hg.1 |
ENST00000370291.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175448.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175448.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175448.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175448.hg.1 |
ENST00000361211.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175448.hg.1 |
ENST00000370287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175448.hg.1 |
ENST00000370291.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175448.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175448.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175448.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175448.hg.1 |
TC0X00011134.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00175453.hg.1 |
ENST00000370287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175453.hg.1 |
ENST00000370291.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175453.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175453.hg.1 |
ENST00000370287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175453.hg.1 |
ENST00000370291.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175453.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175453.hg.1 |
TC0X00011134.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00175445.hg.1 |
ENST00000361211.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175445.hg.1 |
ENST00000370287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175445.hg.1 |
ENST00000370291.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175445.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175445.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175445.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175445.hg.1 |
ENST00000361211.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175445.hg.1 |
ENST00000370287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175445.hg.1 |
ENST00000370291.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175445.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175445.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175445.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175445.hg.1 |
TC0X00011134.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00175451.hg.1 |
ENST00000361211.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175451.hg.1 |
ENST00000370287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175451.hg.1 |
ENST00000370291.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175451.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175451.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175451.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175451.hg.1 |
ENST00000361211.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175451.hg.1 |
ENST00000370287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175451.hg.1 |
ENST00000370291.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175451.hg.1 |
ENST00000452779.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00175451.hg.1 |
NM_001102576.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175451.hg.1 |
NM_153478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00175451.hg.1 |
TC0X00011134.hg.1 |
Clariom D TC |