Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCUBE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005248943.1 | XP_005249000.1 | 6 - 7 | 1087 | 82 | |
XM_005248947.1 | XP_005249004.1 | 6 - 7 | 1087 | 82 | ||
XM_011514404.2 | XP_011512706.1 | 6 - 7 | 787 | 82 | ||
GenBank mRNA | AK092062.1 | - | 5 - 6 | 583 | 82 | |
CR936607.1 | - | 2 - 3 | 245 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |