Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08002808.hg.1 |
TC08000196.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08002808.hg.1 |
ENST00000330560.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08002808.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08002808.hg.1 |
ENST00000434814.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08002808.hg.1 |
ENST00000523454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08002808.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000518225.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000518225.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000521098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000521098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000521098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000521098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000521098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000521098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000521098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000521098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000521098.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000523454.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000523454.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000523454.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000523454.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000523454.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000523454.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
ENST00000523454.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08002811.hg.1 |
TC08000196.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08002811.hg.1 |
ENST00000330560.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08002811.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08002811.hg.1 |
ENST00000434814.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08002811.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148433.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148433.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148433.hg.1 |
ENST00000521098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148433.hg.1 |
ENST00000523454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148433.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148433.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148433.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148433.hg.1 |
ENST00000521098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148433.hg.1 |
ENST00000523454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148433.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148433.hg.1 |
TC0800007065.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000330560.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000380665.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000518225.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000518225.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000521098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000521098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000521098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000521098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000521098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000521098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000521098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000521098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000521098.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000523454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000523454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000523454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000523454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000523454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000523454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
ENST00000523454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007065.hg.1 |
NM_152562.3 |
Refseq |