Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000332220.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000332220.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000332220.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000332220.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000332220.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000332220.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000332220.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000332220.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000467957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000467957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000467957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000467957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000467957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000467957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000467957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012164.hg.1 |
ENST00000489612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012164.hg.1 |
ENST00000489612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012164.hg.1 |
ENST00000489612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156581.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156581.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156581.hg.1 |
ENST00000467957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156581.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156581.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100156581.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156581.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156581.hg.1 |
ENST00000467957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156581.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156581.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100156581.hg.1 |
TC0100008025.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700012164.hg.1 |
ENST00000489612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012164.hg.1 |
ENST00000489612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012164.hg.1 |
ENST00000489612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000321358.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000332220.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000332220.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000332220.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000332220.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000332220.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000332220.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000332220.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000332220.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000436427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000467957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000467957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000467957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000467957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000467957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000467957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000467957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
ENST00000616651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008025.hg.1 |
NM_004559.4 |
Refseq |