Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000356300.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000356300.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000356300.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000356300.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000356300.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000356300.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000356300.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000356300.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000356300.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000356300.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000397118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000397118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000397118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000397118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000397118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000397118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000397118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000397118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000397118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000397118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000397118.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000544177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000544177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000544177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000544177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000544177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000553505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000553505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000553505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000553505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000553505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000622501.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000622501.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000622501.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000622501.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000622501.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000622501.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
ENST00000622501.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
NM_001146028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
NM_001146028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
NM_001146028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
NM_001146028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
NM_001146028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
NM_001146028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
NM_001146028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
NM_001146028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
NM_001146028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
NM_001146028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
NM_032452.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
NM_032452.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
NM_032452.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
NM_032452.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
NM_032452.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
NM_032452.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
NM_032452.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
NM_032452.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
NM_032452.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
NM_032452.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010719.hg.1 |
NM_032452.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010804.hg.1 |
TC14002206.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010804.hg.1 |
ENST00000267407.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010804.hg.1 |
ENST00000356300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010804.hg.1 |
ENST00000397118.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010804.hg.1 |
ENST00000543864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010804.hg.1 |
ENST00000544177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010804.hg.1 |
NM_001146028.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010804.hg.1 |
NM_032452.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400181570.hg.1 |
ENST00000356300.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181570.hg.1 |
ENST00000397118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181570.hg.1 |
ENST00000544177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181570.hg.1 |
ENST00000622501.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181570.hg.1 |
NM_001146028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400181570.hg.1 |
NM_032452.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400181570.hg.1 |
ENST00000356300.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181570.hg.1 |
ENST00000397118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181570.hg.1 |
ENST00000544177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181570.hg.1 |
ENST00000622501.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400181570.hg.1 |
NM_001146028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400181570.hg.1 |
NM_032452.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400181570.hg.1 |
TC1400010719.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000356300.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000356300.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000356300.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000356300.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000356300.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000356300.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000356300.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000356300.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000356300.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000356300.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000397118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000397118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000397118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000397118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000397118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000397118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000397118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000397118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000397118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000397118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000397118.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000544177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000544177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000544177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000544177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000544177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000553505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000553505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000553505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000553505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000553505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000622501.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000622501.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000622501.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000622501.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000622501.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000622501.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
ENST00000622501.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
NM_001146028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
NM_001146028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
NM_001146028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
NM_001146028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
NM_001146028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
NM_001146028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
NM_001146028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
NM_001146028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
NM_001146028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
NM_001146028.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
NM_032452.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
NM_032452.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
NM_032452.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
NM_032452.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
NM_032452.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
NM_032452.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
NM_032452.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
NM_032452.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
NM_032452.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
NM_032452.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010719.hg.1 |
NM_032452.2 |
Refseq |