Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052429.hg.1 |
TC01003355.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052429.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052429.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016025.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221657.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221657.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221657.hg.1 |
ENST00000368195.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221657.hg.1 |
NM_014215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221657.hg.1 |
TC0100016025.hg.1 |
Clariom D TC |