Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAJ17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC035680.1 | - | 1 - 2 | 226 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons |