Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012220.hg.1 |
ENST00000504237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012220.hg.1 |
ENST00000504237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012220.hg.1 |
ENST00000504237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012220.hg.1 |
ENST00000511399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012220.hg.1 |
ENST00000511399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012220.hg.1 |
ENST00000511399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012220.hg.1 |
ENST00000511399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012220.hg.1 |
ENST00000511399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012220.hg.1 |
ENST00000504237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012220.hg.1 |
ENST00000504237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012220.hg.1 |
ENST00000504237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012220.hg.1 |
ENST00000511399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012220.hg.1 |
ENST00000511399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012220.hg.1 |
ENST00000511399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012220.hg.1 |
ENST00000511399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012220.hg.1 |
ENST00000511399.1 |
Ensembl |