Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011255.hg.1 |
ENST00000399966.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011255.hg.1 |
ENST00000399966.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011255.hg.1 |
ENST00000399966.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011255.hg.1 |
ENST00000399966.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007317.hg.1 |
ENSTR0000399966.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007317.hg.1 |
ENSTR0000399966.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007317.hg.1 |
ENSTR0000399966.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007317.hg.1 |
ENSTR0000399966.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006433.hg.1 |
ENST00000329244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006433.hg.1 |
ENST00000329244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006433.hg.1 |
ENST00000329244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006433.hg.1 |
ENST00000329244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006433.hg.1 |
ENST00000329244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006433.hg.1 |
ENST00000527434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006433.hg.1 |
ENST00000527434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006433.hg.1 |
ENST00000527434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006433.hg.1 |
ENST00000527434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006433.hg.1 |
NR_130735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006433.hg.1 |
NR_130735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006433.hg.1 |
NR_130735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006433.hg.1 |
NR_130735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144064.hg.1 |
ENST00000329244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144064.hg.1 |
ENST00000527434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144064.hg.1 |
NR_130735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144064.hg.1 |
ENST00000329244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144064.hg.1 |
ENST00000527434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144064.hg.1 |
NR_130735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144064.hg.1 |
TC1600006433.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600144059.hg.1 |
ENST00000329244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144059.hg.1 |
ENST00000527434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144059.hg.1 |
NR_130735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144059.hg.1 |
ENST00000329244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144059.hg.1 |
ENST00000527434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144059.hg.1 |
NR_130735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144059.hg.1 |
TC1600006433.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0Y00007317.hg.1 |
ENSTR0000399966.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007317.hg.1 |
ENSTR0000399966.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007317.hg.1 |
ENSTR0000399966.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007317.hg.1 |
ENSTR0000399966.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144063.hg.1 |
ENST00000329244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144063.hg.1 |
ENST00000527434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144063.hg.1 |
NR_130735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144063.hg.1 |
ENST00000329244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144063.hg.1 |
ENST00000527434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144063.hg.1 |
NR_130735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144063.hg.1 |
TC1600006433.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600144062.hg.1 |
ENST00000329244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144062.hg.1 |
ENST00000527434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144062.hg.1 |
NR_130735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144062.hg.1 |
ENST00000329244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144062.hg.1 |
ENST00000527434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144062.hg.1 |
NR_130735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144062.hg.1 |
TC1600006433.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00011255.hg.1 |
ENST00000399966.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011255.hg.1 |
ENST00000399966.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011255.hg.1 |
ENST00000399966.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011255.hg.1 |
ENST00000399966.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006433.hg.1 |
ENST00000329244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006433.hg.1 |
ENST00000329244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006433.hg.1 |
ENST00000329244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006433.hg.1 |
ENST00000329244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006433.hg.1 |
ENST00000329244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006433.hg.1 |
ENST00000527434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006433.hg.1 |
ENST00000527434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006433.hg.1 |
ENST00000527434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006433.hg.1 |
ENST00000527434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006433.hg.1 |
NR_130735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006433.hg.1 |
NR_130735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006433.hg.1 |
NR_130735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006433.hg.1 |
NR_130735.1 |
Refseq |