Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.1 ST Array |
8151125 |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010696.hg.1 |
ENST00000563496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010696.hg.1 |
ENST00000563496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010696.hg.1 |
ENST00000563496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010696.hg.1 |
ENST00000563496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010696.hg.1 |
ENST00000563496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010696.hg.1 |
ENST00000563496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010696.hg.1 |
ENST00000566129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010696.hg.1 |
ENST00000566129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010696.hg.1 |
ENST00000566129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010696.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010696.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010696.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010696.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010696.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010696.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016596.hg.1 |
TC08001291.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016596.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016597.hg.1 |
TC08001291.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016597.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016598.hg.1 |
TC08001291.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016598.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016599.hg.1 |
TC08001291.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016599.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172599.hg.1 |
ENST00000563496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172599.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172599.hg.1 |
ENST00000563496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172599.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172599.hg.1 |
TC0800010696.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800172594.hg.1 |
ENST00000563496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172594.hg.1 |
ENST00000566129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172594.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172594.hg.1 |
ENST00000563496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172594.hg.1 |
ENST00000566129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172594.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172594.hg.1 |
TC0800010696.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800172598.hg.1 |
ENST00000563496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172598.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172598.hg.1 |
ENST00000563496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172598.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172598.hg.1 |
TC0800010696.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800010696.hg.1 |
ENST00000563496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010696.hg.1 |
ENST00000563496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010696.hg.1 |
ENST00000563496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010696.hg.1 |
ENST00000563496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010696.hg.1 |
ENST00000563496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010696.hg.1 |
ENST00000563496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010696.hg.1 |
ENST00000566129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010696.hg.1 |
ENST00000566129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010696.hg.1 |
ENST00000566129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010696.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010696.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010696.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010696.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010696.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010696.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172596.hg.1 |
ENST00000563496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172596.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172596.hg.1 |
ENST00000563496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172596.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172596.hg.1 |
TC0800010696.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800172593.hg.1 |
ENST00000563496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172593.hg.1 |
ENST00000566129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172593.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172593.hg.1 |
ENST00000563496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172593.hg.1 |
ENST00000566129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172593.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172593.hg.1 |
TC0800010696.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800172592.hg.1 |
ENST00000563496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172592.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172592.hg.1 |
ENST00000563496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172592.hg.1 |
NM_001193502.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172592.hg.1 |
TC0800010696.hg.1 |
Clariom D TC |