Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011061.hg.1 |
TC08000836.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011061.hg.1 |
ENST00000377470.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011061.hg.1 |
ENST00000447830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011061.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011061.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011064.hg.1 |
TC08000836.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011064.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011064.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
ENST00000377470.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
ENST00000377470.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
ENST00000377470.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
ENST00000377470.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
ENST00000377470.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
ENST00000377470.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
ENST00000377470.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
ENST00000377470.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
ENST00000447830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
ENST00000447830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
ENST00000447830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
ENST00000447830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
ENST00000447830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
ENST00000447830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009232.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162971.hg.1 |
ENST00000377470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800162971.hg.1 |
ENST00000447830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800162971.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162971.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162971.hg.1 |
ENST00000377470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800162971.hg.1 |
ENST00000447830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800162971.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162971.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162971.hg.1 |
TC0800009232.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
ENST00000377470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
ENST00000377470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
ENST00000377470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
ENST00000377470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
ENST00000377470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
ENST00000377470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
ENST00000377470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
ENST00000377470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
ENST00000447830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
ENST00000447830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
ENST00000447830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
ENST00000447830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
ENST00000447830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
ENST00000447830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009232.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162974.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162974.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162974.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162974.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162974.hg.1 |
TC0800009232.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800162970.hg.1 |
ENST00000377470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800162970.hg.1 |
ENST00000447830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800162970.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162970.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162970.hg.1 |
ENST00000377470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800162970.hg.1 |
ENST00000447830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800162970.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162970.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162970.hg.1 |
TC0800009232.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800162973.hg.1 |
ENST00000377470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800162973.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162973.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162973.hg.1 |
ENST00000377470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800162973.hg.1 |
NM_001134374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162973.hg.1 |
NM_198572.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162973.hg.1 |
TC0800009232.hg.1 |
Clariom D TC |