Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000502576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000502576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000502576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000502576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000502576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000502576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000507286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000507286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000507286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000507286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000507286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000507286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000507286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000507518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000507518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000507518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000507518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000507518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000507518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000507518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000507518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000508413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000508413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000508413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000508413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000512344.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000512344.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000512344.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000512344.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000512344.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000512344.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000513854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000513854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000513854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000513854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
ENST00000513854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000358290.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000502576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000502576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000502576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000502576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000502576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000502576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000507286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000507286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000507286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000507286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000507286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000507286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000507286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000507518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000507518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000507518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000507518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000507518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000507518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000507518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000507518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000508413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000508413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000508413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000508413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000512344.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000512344.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000512344.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000512344.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000512344.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000512344.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000513854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000513854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000513854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000513854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
ENST00000513854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012945.hg.1 |
NM_016245.4 |
Refseq |