Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006623.hg.1 |
ENST00000419844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000261813.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000261813.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000261813.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000261813.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000261813.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000261813.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000261813.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000261813.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000261813.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000261813.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000507185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000507185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000507185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000507185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000510217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000510217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000510217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000510217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000510217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000510217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000510217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000512707.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000512707.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000512707.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000512707.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000512707.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000512925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000512925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000512925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000512925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000514611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000514611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000514611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000514611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000514611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000514611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000514611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000524074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000524074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000524074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
ENST00000524074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012236.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05025429.hg.1 |
TC05001853.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05025429.hg.1 |
ENST00000261813.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05025429.hg.1 |
ENST00000510217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05025429.hg.1 |
ENST00000524074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05025429.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05025427.hg.1 |
TC05001853.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05025427.hg.1 |
ENST00000261813.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05025427.hg.1 |
ENST00000514611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05025427.hg.1 |
ENST00000524074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05025427.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500184939.hg.1 |
ENST00000261813.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184939.hg.1 |
ENST00000507185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184939.hg.1 |
ENST00000514611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184939.hg.1 |
ENST00000524074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184939.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500184939.hg.1 |
ENST00000261813.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184939.hg.1 |
ENST00000507185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184939.hg.1 |
ENST00000514611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184939.hg.1 |
ENST00000524074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184939.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500184939.hg.1 |
TC0500012236.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300006623.hg.1 |
ENST00000419844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000261813.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000261813.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000261813.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000261813.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000261813.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000261813.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000261813.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000261813.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000261813.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000261813.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000507185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000507185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000507185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000507185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000510217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000510217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000510217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000510217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000510217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000510217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000510217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000512707.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000512707.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000512707.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000512707.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000512707.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000512925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000512925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000512925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000512925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000514611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000514611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000514611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000514611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000514611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000514611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000514611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000524074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000524074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000524074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
ENST00000524074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012236.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500184941.hg.1 |
ENST00000261813.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184941.hg.1 |
ENST00000507185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184941.hg.1 |
ENST00000510217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184941.hg.1 |
ENST00000524074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184941.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500184941.hg.1 |
ENST00000261813.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184941.hg.1 |
ENST00000507185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184941.hg.1 |
ENST00000510217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184941.hg.1 |
ENST00000524074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184941.hg.1 |
NM_002622.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500184941.hg.1 |
TC0500012236.hg.1 |
Clariom D TC |