Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007964.hg.1 |
ENST00000334346.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007964.hg.1 |
ENST00000334346.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007964.hg.1 |
ENST00000334346.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007964.hg.1 |
ENST00000334346.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007964.hg.1 |
ENST00000334346.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007964.hg.1 |
ENST00000562399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007964.hg.1 |
ENST00000562399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007964.hg.1 |
ENST00000562399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007964.hg.1 |
ENST00000562399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007964.hg.1 |
ENST00000562399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007964.hg.1 |
ENST00000567054.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007964.hg.1 |
ENST00000567054.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007964.hg.1 |
ENST00000568608.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007964.hg.1 |
ENST00000568608.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007964.hg.1 |
ENST00000568608.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007964.hg.1 |
NM_005947.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007964.hg.1 |
NM_005947.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007964.hg.1 |
NM_005947.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007964.hg.1 |
NM_005947.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007964.hg.1 |
NM_005947.2 |
Refseq |