Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000588526.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000588526.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000588526.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000588526.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000588526.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000588526.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000588526.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000588526.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000589290.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000589290.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000589290.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000589290.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000593245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000593245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000593245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000593245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000593245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000593245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
ENST00000593245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011674.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003531.hg.1 |
ENST00000040663.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003531.hg.1 |
ENST00000319545.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003531.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003531.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003531.hg.1 |
TC19000239.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003532.hg.1 |
ENST00000040663.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003532.hg.1 |
ENST00000319545.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003532.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003532.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003532.hg.1 |
TC19000239.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003533.hg.1 |
ENST00000040663.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003533.hg.1 |
ENST00000319545.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003533.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003533.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003533.hg.1 |
TC19000239.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900199538.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199538.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199538.hg.1 |
ENST00000588526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199538.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199538.hg.1 |
ENST00000593245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199538.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199538.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199538.hg.1 |
TC1900011674.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900199538.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199538.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199538.hg.1 |
ENST00000588526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199538.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199538.hg.1 |
ENST00000593245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199538.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199538.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199540.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199540.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199540.hg.1 |
ENST00000588526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199540.hg.1 |
ENST00000589290.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199540.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199540.hg.1 |
ENST00000593245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199540.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199540.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199540.hg.1 |
TC1900011674.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900199540.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199540.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199540.hg.1 |
ENST00000588526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199540.hg.1 |
ENST00000589290.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199540.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199540.hg.1 |
ENST00000593245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199540.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199540.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000588526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000588526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000588526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000588526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000588526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000588526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000588526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000588526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000589290.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000589290.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000589290.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000589290.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000593245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000593245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000593245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000593245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000593245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000593245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
ENST00000593245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011674.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199541.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199541.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199541.hg.1 |
ENST00000588526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199541.hg.1 |
ENST00000589290.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199541.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199541.hg.1 |
ENST00000593245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199541.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199541.hg.1 |
NM_032285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199541.hg.1 |
TC1900011674.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900199541.hg.1 |
ENST00000040663.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199541.hg.1 |
ENST00000319545.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199541.hg.1 |
ENST00000588526.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199541.hg.1 |
ENST00000589290.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199541.hg.1 |
ENST00000591688.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199541.hg.1 |
ENST00000593245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199541.hg.1 |
NM_001031727.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199541.hg.1 |
NM_032285.3 |
Refseq |