Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010924.hg.1 |
TC02000706.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010924.hg.1 |
ENST00000233336.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010924.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
ENST00000460450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
ENST00000460450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
ENST00000460450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
ENST00000460450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
ENST00000476615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
ENST00000476615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
ENST00000476615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
ENST00000476615.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
ENST00000485537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
ENST00000485537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
ENST00000485537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162252.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162252.hg.1 |
ENST00000476615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162252.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162252.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162252.hg.1 |
ENST00000476615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162252.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162252.hg.1 |
TC0200008921.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
ENST00000233336.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
ENST00000460450.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
ENST00000460450.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
ENST00000460450.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
ENST00000460450.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
ENST00000476615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
ENST00000476615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
ENST00000476615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
ENST00000476615.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
ENST00000485537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
ENST00000485537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
ENST00000485537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008921.hg.1 |
NM_153712.4 |
Refseq |