Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017197.hg.1 |
TC01001092.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017197.hg.1 |
ENST00000369308.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017197.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017199.hg.1 |
TC01001092.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017199.hg.1 |
ENST00000369308.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017199.hg.1 |
ENST00000449167.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017199.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017196.hg.1 |
TC01001092.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017196.hg.1 |
ENST00000369308.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017196.hg.1 |
ENST00000449167.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017196.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017195.hg.1 |
TC01001092.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017195.hg.1 |
ENST00000369308.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017195.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216650.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216650.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216650.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216650.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216650.hg.1 |
TC0100015596.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216649.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216649.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216649.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216649.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216649.hg.1 |
TC0100015596.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100216651.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216651.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216651.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216651.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216651.hg.1 |
TC0100015596.hg.1 |
Clariom D TC |