Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNQ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172107.3 | NP_742105.1 | 12 - 13 | 1515 | 71 | |
XM_017027845.1 | XP_016883334.1 | 1 - 2 | 813 | 71 | ||
GenBank mRNA | AF033348.1 | - | 12 - 13 | 1428 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Homo sapiens Probe spans exons |