Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009522.hg.1 |
TC04000653.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009522.hg.1 |
ENST00000281146.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009522.hg.1 |
ENST00000425929.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009522.hg.1 |
ENST00000502887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009522.hg.1 |
NM_001099783.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009522.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000425929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000425929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000425929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000425929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000425929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000425929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000425929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000425929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000425929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000502360.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000502360.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000502360.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000502360.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000502887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000502887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000502887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000502887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000502887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000502887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000502887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000502887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000508622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000508622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000508622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000508622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000508622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000508673.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000508673.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000508673.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000508673.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
ENST00000508673.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
NM_001099783.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
NM_001099783.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
NM_001099783.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
NM_001099783.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
NM_001099783.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
NM_001099783.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
NM_001099783.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
NM_001099783.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
NM_001099783.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008686.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009521.hg.1 |
TC04000653.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009521.hg.1 |
ENST00000281146.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009521.hg.1 |
ENST00000425929.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009521.hg.1 |
ENST00000502887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009521.hg.1 |
NM_001099783.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009521.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000425929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000425929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000425929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000425929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000425929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000425929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000425929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000425929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000425929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000502360.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000502360.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000502360.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000502360.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000502887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000502887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000502887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000502887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000502887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000502887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000502887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000502887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000508622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000508622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000508622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000508622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000508622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000508673.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000508673.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000508673.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000508673.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
ENST00000508673.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
NM_001099783.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
NM_001099783.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
NM_001099783.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
NM_001099783.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
NM_001099783.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
NM_001099783.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
NM_001099783.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
NM_001099783.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
NM_001099783.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008686.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400159949.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400159949.hg.1 |
ENST00000425929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400159949.hg.1 |
ENST00000502887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400159949.hg.1 |
ENST00000508673.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400159949.hg.1 |
NM_001099783.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400159949.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400159949.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400159949.hg.1 |
ENST00000425929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400159949.hg.1 |
ENST00000502887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400159949.hg.1 |
ENST00000508673.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400159949.hg.1 |
NM_001099783.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400159949.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400159949.hg.1 |
TC0400008686.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400159950.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400159950.hg.1 |
ENST00000425929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400159950.hg.1 |
ENST00000502887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400159950.hg.1 |
NM_001099783.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400159950.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400159950.hg.1 |
ENST00000281146.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400159950.hg.1 |
ENST00000425929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400159950.hg.1 |
ENST00000502887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400159950.hg.1 |
NM_001099783.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400159950.hg.1 |
NM_173487.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400159950.hg.1 |
TC0400008686.hg.1 |
Clariom D TC |