Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015582.hg.1 |
ENST00000329451.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015582.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015582.hg.1 |
TC11001237.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015580.hg.1 |
ENST00000329451.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015580.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015580.hg.1 |
TC11001237.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000441853.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000441853.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000441853.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000441853.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000441853.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000441853.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000441853.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000441853.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000441853.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000441853.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000469990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000469990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000469990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000469990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000469990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000469990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000469990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000469990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000486629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000486629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000486629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000486629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000486629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000486629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000486629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000486629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000492515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000492515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000492515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
ENST00000492515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100172888.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172888.hg.1 |
ENST00000469990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172888.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100172888.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172888.hg.1 |
ENST00000469990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172888.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100172888.hg.1 |
TC1100009679.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000441853.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000441853.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000441853.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000441853.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000441853.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000441853.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000441853.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000441853.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000441853.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000441853.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000469990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000469990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000469990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000469990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000469990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000469990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000469990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000469990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000486629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000486629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000486629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000486629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000486629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000486629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000486629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000486629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000492515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000492515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000492515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
ENST00000492515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009679.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100172886.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172886.hg.1 |
ENST00000469990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172886.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100172886.hg.1 |
ENST00000329451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172886.hg.1 |
ENST00000469990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172886.hg.1 |
NM_173573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100172886.hg.1 |
TC1100009679.hg.1 |
Clariom D TC |