Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15008493.hg.1 |
ENST00000268164.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15008493.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15008493.hg.1 |
TC15000883.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15008495.hg.1 |
ENST00000268164.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15008495.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15008495.hg.1 |
TC15000883.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000539113.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000539113.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000539113.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000539113.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000539113.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000539113.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000555434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000555434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000555434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000555434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000555434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000556382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000556382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000556382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000556382.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15008494.hg.1 |
ENST00000268164.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15008494.hg.1 |
ENST00000539113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15008494.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15008494.hg.1 |
TC15000883.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500160701.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500160701.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500160701.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500160701.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500160701.hg.1 |
TC1500008370.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500160703.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500160703.hg.1 |
ENST00000539113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500160703.hg.1 |
ENST00000555434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500160703.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500160703.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500160703.hg.1 |
ENST00000539113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500160703.hg.1 |
ENST00000555434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500160703.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500160703.hg.1 |
TC1500008370.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500160704.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500160704.hg.1 |
ENST00000555434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500160704.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500160704.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500160704.hg.1 |
ENST00000555434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500160704.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500160704.hg.1 |
TC1500008370.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000539113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000539113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000539113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000539113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000539113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000539113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000555434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000555434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000555434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000555434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000555434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000556382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000556382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000556382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000556382.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |