Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
ENST00000341010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
ENST00000341010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
ENST00000341010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
ENST00000341010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
ENST00000341010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
ENST00000341010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
ENST00000341010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
ENST00000341010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
ENST00000341010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
ENST00000393197.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
ENST00000393197.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
ENST00000393197.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
ENST00000393197.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
ENST00000393197.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
ENST00000393197.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
ENST00000393197.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
ENST00000393197.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
ENST00000393197.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
ENST00000496265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
ENST00000496265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
ENST00000496265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
ENST00000496265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
ENST00000496265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
NM_032556.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
NM_032556.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
NM_032556.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
NM_032556.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
NM_032556.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
NM_032556.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
NM_032556.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
NM_032556.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
NM_032556.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
NM_173161.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
NM_173161.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
NM_173161.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
NM_173161.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
NM_173161.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
NM_173161.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
NM_173161.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
NM_173161.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008950.hg.1 |
NM_173161.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011134.hg.1 |
TC02000718.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011134.hg.1 |
ENST00000337569.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011134.hg.1 |
ENST00000341010.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011134.hg.1 |
ENST00000393197.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011134.hg.1 |
NM_032556.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011134.hg.1 |
NM_173161.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162501.hg.1 |
ENST00000341010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162501.hg.1 |
ENST00000393197.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162501.hg.1 |
ENST00000496265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162501.hg.1 |
NM_032556.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162501.hg.1 |
NM_173161.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162501.hg.1 |
ENST00000341010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162501.hg.1 |
ENST00000393197.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162501.hg.1 |
ENST00000496265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200162501.hg.1 |
NM_032556.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162501.hg.1 |
NM_173161.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200162501.hg.1 |
TC0200008950.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
ENST00000341010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
ENST00000341010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
ENST00000341010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
ENST00000341010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
ENST00000341010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
ENST00000341010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
ENST00000341010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
ENST00000341010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
ENST00000341010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
ENST00000393197.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
ENST00000393197.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
ENST00000393197.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
ENST00000393197.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
ENST00000393197.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
ENST00000393197.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
ENST00000393197.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
ENST00000393197.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
ENST00000393197.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
ENST00000496265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
ENST00000496265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
ENST00000496265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
ENST00000496265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
ENST00000496265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
NM_032556.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
NM_032556.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
NM_032556.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
NM_032556.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
NM_032556.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
NM_032556.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
NM_032556.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
NM_032556.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
NM_032556.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
NM_173161.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
NM_173161.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
NM_173161.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
NM_173161.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
NM_173161.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
NM_173161.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
NM_173161.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
NM_173161.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008950.hg.1 |
NM_173161.2 |
Refseq |