Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
NM_001898_at |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8065414 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8065413 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8065415 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8065412 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
206224_PM_at |
- |
- |
Affymetrix Human Genome U219 Array |
11730185_x_at |
- |
- |
Affymetrix Human Genome U219 Array |
11730185_PM_x_at |
- |
- |
Affymetrix Human Genome U219 Array |
11730184_at |
- |
- |
Affymetrix Human Genome U219 Array |
11730184_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009763.hg.1 |
ENST00000304749.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009763.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009763.hg.1 |
TC20000718.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008682.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008682.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008682.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008682.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008682.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008682.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008682.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008682.hg.1 |
ENST00000398402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008682.hg.1 |
ENST00000398402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008682.hg.1 |
ENST00000398402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008682.hg.1 |
ENST00000398402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008682.hg.1 |
ENST00000398402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008682.hg.1 |
ENST00000398402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008682.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008682.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008682.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008682.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008682.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008682.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008682.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008680.hg.1 |
ENST00000217423.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008680.hg.1 |
ENST00000217423.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008680.hg.1 |
ENST00000217423.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008680.hg.1 |
ENST00000217423.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008680.hg.1 |
ENST00000217423.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008680.hg.1 |
ENST00000217423.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008680.hg.1 |
ENST00000217423.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008680.hg.1 |
NM_001899.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008680.hg.1 |
NM_001899.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008680.hg.1 |
NM_001899.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008680.hg.1 |
NM_001899.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008680.hg.1 |
NM_001899.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008680.hg.1 |
NM_001899.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008680.hg.1 |
NM_001899.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008684.hg.1 |
ENST00000304725.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008684.hg.1 |
ENST00000304725.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008684.hg.1 |
ENST00000304725.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008684.hg.1 |
ENST00000304725.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008684.hg.1 |
ENST00000304725.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008684.hg.1 |
NM_001322.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008684.hg.1 |
NM_001322.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008684.hg.1 |
NM_001322.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008684.hg.1 |
NM_001322.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009764.hg.1 |
ENST00000304749.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009764.hg.1 |
ENST00000398402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009764.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009764.hg.1 |
TC20000718.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009767.hg.1 |
ENST00000304749.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009767.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009767.hg.1 |
TC20000718.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009765.hg.1 |
ENST00000304749.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009765.hg.1 |
ENST00000398402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009765.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009765.hg.1 |
TC20000718.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009766.hg.1 |
ENST00000304749.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009766.hg.1 |
ENST00000398402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009766.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009766.hg.1 |
TC20000718.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000160774.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160774.hg.1 |
ENST00000398402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160774.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160774.hg.1 |
TC2000008682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160774.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160774.hg.1 |
ENST00000398402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160774.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008684.hg.1 |
ENST00000304725.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008684.hg.1 |
ENST00000304725.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008684.hg.1 |
ENST00000304725.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008684.hg.1 |
ENST00000304725.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008684.hg.1 |
ENST00000304725.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008684.hg.1 |
NM_001322.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008684.hg.1 |
NM_001322.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008684.hg.1 |
NM_001322.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008684.hg.1 |
NM_001322.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008682.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008682.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008682.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008682.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008682.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008682.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008682.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008682.hg.1 |
ENST00000398402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008682.hg.1 |
ENST00000398402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008682.hg.1 |
ENST00000398402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008682.hg.1 |
ENST00000398402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008682.hg.1 |
ENST00000398402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008682.hg.1 |
ENST00000398402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008682.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008682.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008682.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008682.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008682.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008682.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008682.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160772.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160772.hg.1 |
ENST00000398402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160772.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160772.hg.1 |
TC2000008682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160772.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160772.hg.1 |
ENST00000398402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160772.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160773.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160773.hg.1 |
ENST00000398402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160773.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160773.hg.1 |
TC2000008682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160773.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160773.hg.1 |
ENST00000398402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160773.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160775.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160775.hg.1 |
ENST00000398402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160775.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160775.hg.1 |
TC2000008682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160775.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160775.hg.1 |
ENST00000398402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160775.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160776.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160776.hg.1 |
ENST00000398402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160776.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160776.hg.1 |
TC2000008682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160776.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160776.hg.1 |
ENST00000398402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160776.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008680.hg.1 |
ENST00000217423.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008680.hg.1 |
ENST00000217423.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008680.hg.1 |
ENST00000217423.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008680.hg.1 |
ENST00000217423.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008680.hg.1 |
ENST00000217423.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008680.hg.1 |
ENST00000217423.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008680.hg.1 |
ENST00000217423.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008680.hg.1 |
NM_001899.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008680.hg.1 |
NM_001899.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008680.hg.1 |
NM_001899.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008680.hg.1 |
NM_001899.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008680.hg.1 |
NM_001899.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008680.hg.1 |
NM_001899.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008680.hg.1 |
NM_001899.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160771.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160771.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160771.hg.1 |
TC2000008682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160771.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160771.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160777.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160777.hg.1 |
NM_001898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160777.hg.1 |
TC2000008682.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160777.hg.1 |
ENST00000304749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160777.hg.1 |
NM_001898.2 |
Refseq |
Human Genome U133 Plus 2.0 Array |
206224_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
206224_at |
- |
- |
Human Genome U133A 2.0 Array |
206224_at |
- |
- |
Human Genome U133A 2.0 Array |
206224_PM_at |
- |
- |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) |
NM_001898_at |
- |
- |
AffymetrixPathwork Diagnostics Pathchip |
pw535035 |
- |
- |
Human Genome U95 Set |
35506_PM_s_at |
- |
- |
Human Genome U95 Set |
35506_s_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
206224_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
206224_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
206224_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
206224_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
206224_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
206224_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
206224_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
206224_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
206224_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
206224_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
206224_at10 |
- |
- |
Human Genome U133 Set |
206224_PM_at |
- |
- |
Human Genome U133 Set |
206224_at |
- |
- |