Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004304.4 | NP_004295.2 | 24 - 25 | 4694 | 71 | |
GenBank mRNA | AB209477.4 | - | 24 - 25 | 4231 | 71 | |
EU788003.1 | - | 24 - 25 | 4649 | 71 | ||
EU788004.1 | - | 24 - 25 | 4649 | 71 | ||
EU788005.1 | - | 24 - 25 | 4649 | 71 | ||
EU788006.1 | - | 24 - 25 | 4649 | 71 | ||
EU788007.1 | - | 24 - 25 | 4649 | 71 | ||
EU788008.1 | - | 24 - 25 | 4649 | 71 | ||
EU788009.1 | - | 24 - 25 | 71 | |||
EU788010.1 | - | 24 - 25 | 4649 | 71 | ||
FJ838786.1 | - | 24 - 25 | 4649 | 71 | ||
FJ838787.1 | - | 24 - 25 | 4649 | 71 | ||
GU128152.1 | - | 24 - 25 | 3742 | 71 | ||
GU128153.1 | - | 24 - 25 | 3742 | 71 | ||
GU128154.1 | - | 24 - 25 | 3742 | 71 | ||
GU128155.1 | - | 24 - 25 | 3742 | 71 | ||
GU128156.1 | - | 24 - 25 | 3742 | 71 | ||
GU128157.1 | - | 24 - 25 | 71 | |||
LN864494.1 | - | 5 - 6 | 942 | 71 | ||
U62540.1 | - | 24 - 25 | 4653 | 71 | ||
U66559.1 | - | 24 - 25 | 3890 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Homo sapiens Human Protein Kinase Pathways Inventoried Human_Protein_Kinase Probe spans exons |