Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001594.hg.1 |
TC01000099.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001594.hg.1 |
ENST00000054666.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001594.hg.1 |
ENST00000470357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01001594.hg.1 |
NM_004781.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000470357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000470357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000470357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000470357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000470357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000487194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000487194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
ENST00000487194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146557.hg.1 |
ENST00000054666.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146557.hg.1 |
ENST00000470357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146557.hg.1 |
ENST00000487194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146557.hg.1 |
NM_004781.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146557.hg.1 |
ENST00000054666.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146557.hg.1 |
ENST00000470357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146557.hg.1 |
ENST00000487194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100146557.hg.1 |
NM_004781.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100146557.hg.1 |
TC0100006723.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000054666.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000470357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000470357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000470357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000470357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000470357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000487194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000487194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
ENST00000487194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006723.hg.1 |
NM_004781.3 |
Refseq |