Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000561525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000561525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000561525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000561525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000561525.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000561819.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000561819.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000561819.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000561819.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000561819.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000561819.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000564942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000564942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000564942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000564942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000564942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000564942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000564942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000569324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000569324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000569324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000569324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
ENST00000569324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006013.hg.1 |
ENST00000285767.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006013.hg.1 |
ENST00000299192.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006013.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006013.hg.1 |
TC16000436.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600156805.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600156805.hg.1 |
ENST00000561525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600156805.hg.1 |
ENST00000564942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600156805.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600156805.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600156805.hg.1 |
ENST00000561525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600156805.hg.1 |
ENST00000564942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600156805.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600156805.hg.1 |
TC1600007806.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000299192.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000561525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000561525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000561525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000561525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000561525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000561819.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000561819.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000561819.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000561819.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000561819.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000561819.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000564942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000564942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000564942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000564942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000564942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000564942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000564942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000569324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000569324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000569324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000569324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
ENST00000569324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007806.hg.1 |
NM_182922.3 |
Refseq |