Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000481868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000481868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000481868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000481868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000481868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000481868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
ENST00000506969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007628.hg.1 |
TC0X000575.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007628.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007628.hg.1 |
ENST00000371666.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007628.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007629.hg.1 |
TC0X000575.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007629.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007629.hg.1 |
ENST00000371666.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007629.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156206.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156206.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156206.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156206.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156206.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156206.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156206.hg.1 |
TC0X00008230.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00156212.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156212.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156212.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156212.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156212.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156212.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156212.hg.1 |
TC0X00008230.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00156205.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156205.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156205.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156205.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156205.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156205.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156205.hg.1 |
TC0X00008230.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000371642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000371666.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000481868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000481868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000481868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000481868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000481868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000481868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
ENST00000506969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008230.hg.1 |
NM_001560.2 |
Refseq |