Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003790.2 | NP_003781.1 | 8 - 9 | 833 | 96 | |
NM_148965.1 | NP_683866.1 | 8 - 9 | 860 | 96 | ||
NM_148967.1 | NP_683868.1 | 7 - 8 | 698 | 96 | ||
GenBank mRNA | AF026070.1 | - | 8 - 9 | 840 | 96 | |
AF026071.1 | - | 7 - 8 | 934 | 96 | ||
AK094463.1 | - | 2 - 3 | 799 | 96 | ||
AY358309.1 | - | 8 - 9 | 847 | 96 | ||
BC117189.1 | - | 8 - 9 | 813 | 40125740 | 96 | |
U72763.1 | - | 8 - 9 | 745 | 96 | ||
U74611.1 | - | 8 - 9 | 833 | 96 | ||
U75380.1 | - | 8 - 9 | 736 | 96 | ||
U78029.1 | - | 8 - 9 | 745 | 96 | ||
U83597.1 | - | 8 - 9 | 729 | 96 | ||
U94501.1 | - | 8 - 9 | 745 | 96 | ||
U94502.1 | - | 8 - 9 | 748 | 96 | ||
U94503.1 | - | 7 - 8 | 846 | 96 | ||
U94504.1 | - | 7 - 8 | 689 | 96 | ||
U94509.1 | - | 7 - 8 | 610 | 96 | ||
Y09392.1 | - | 7 - 8 | 914 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |