Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP8B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001178002.2 | NP_001171473.1 | 21 - 22 | 2529 | 95 | |
NM_138813.3 | NP_620168.1 | 21 - 22 | 2620 | 95 | ||
NR_047593.2 | 21 - 22 | 2877 | 95 | |||
XM_006722654.2 | XP_006722717.1 | 21 - 22 | 2650 | 95 | ||
XM_006722655.2 | XP_006722718.1 | 21 - 22 | 2729 | 95 | ||
XM_006722656.2 | XP_006722719.1 | 21 - 22 | 2538 | 95 | ||
XM_011527707.1 | XP_011526009.1 | 21 - 22 | 2650 | 95 | ||
XM_011527708.1 | XP_011526010.1 | 21 - 22 | 2530 | 95 | ||
XM_011527710.2 | XP_011526012.1 | 15 - 16 | 1859 | 95 | ||
XM_011527711.2 | XP_011526013.1 | 21 - 22 | 2650 | 95 | ||
XM_011527712.1 | XP_011526014.1 | 6 - 7 | 652 | 95 | ||
GenBank mRNA | AK125969.1 | - | 21 - 22 | 2877 | 95 | |
AY302538.1 | - | 21 - 22 | 2492 | 95 | ||
BC035162.2 | - | 21 - 22 | 2615 | 5265541 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons |