Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000594696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000594696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000594696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000594696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000594696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000594696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000594696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000594696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000599698.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000599698.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000599698.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000599698.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000599698.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000599698.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000599698.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000599698.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000600262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000600262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
ENST00000600262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015561.hg.1 |
ENST00000255612.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015561.hg.1 |
ENST00000423345.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015561.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015561.hg.1 |
TC19001133.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900176712.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900176712.hg.1 |
ENST00000594696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900176712.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900176712.hg.1 |
TC1900009546.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900176712.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900176712.hg.1 |
ENST00000594696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900176712.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000423345.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000594696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000594696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000594696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000594696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000594696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000594696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000594696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000594696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000599698.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000599698.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000599698.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000599698.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000599698.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000599698.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000599698.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000599698.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000600262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000600262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
ENST00000600262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009546.hg.1 |
NM_032152.4 |
Refseq |