Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0400188569.hg.1 |
ENST00000312465.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188569.hg.1 |
ENST00000456523.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188569.hg.1 |
ENST00000503413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188569.hg.1 |
NM_001291812.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400188569.hg.1 |
NM_004464.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400188569.hg.1 |
NM_033143.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400188569.hg.1 |
ENST00000312465.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188569.hg.1 |
ENST00000456523.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188569.hg.1 |
ENST00000503413.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400188569.hg.1 |
NM_001291812.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400188569.hg.1 |
NM_004464.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400188569.hg.1 |
NM_033143.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400188569.hg.1 |
TC0400012820.hg.1 |
Clariom D TC |