Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010919.hg.1 |
TC06000932.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010919.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010919.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163623.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163623.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163623.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163623.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163623.hg.1 |
TC0600009261.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
ENST00000368494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009261.hg.1 |
NM_138459.4 |
Refseq |