Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09388140 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29848083 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29848059 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16716200 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16716202 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09388156 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29848053 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16716203 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29848080 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29848085 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29848054 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29848063 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09388141 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09388164 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09388154 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09388148 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09388150 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09388149 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29848077 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29848069 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09388146 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29848050 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29848056 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09388147 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29848052 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09388138 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29848070 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09388157 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29848072 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P09388158 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
6216 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
24222 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
281010 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P99771 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3245539 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_23_P99771 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
23209 |
- |
- |
Agilent Human Genome CGH Microarray 44B |
A_14_P137045 |
- |
- |
Agilent Human Genome CGH Microarray 44A |
A_14_P137045 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
38065 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
5356 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P02944240 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3245539 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P99771 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_33_P3245539 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_23_P99771 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09388140 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09388148 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09388158 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09388149 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29848059 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29848070 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09388150 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29848063 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09388146 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09388154 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29848083 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29848069 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09388164 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09388156 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29848077 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29848080 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09388157 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16716202 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29848063 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29848077 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09388164 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29848080 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09388156 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09388157 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09388149 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09388140 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09388148 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09388154 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29848083 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29848069 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09388158 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29848070 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29848059 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09388150 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16716202 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P09388146 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_23_P99771 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P137045 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09388151 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09388143 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09388156 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09388148 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09388145 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09388163 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09388146 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09388154 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09388138 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09388160 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P09388158 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16716204 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16716205 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P09388158 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16716207 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16716208 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16716209 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P09388157 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16716206 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P09388160 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P09388159 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P137045 |
- |
- |
Agilent Human Genome CGH Microarray 4x44K |
A_14_P137045 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_23_P99771 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
122277 |
AK131024.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
122277 |
AY358196.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
122277 |
BC029535.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
122277 |
NM_152358.2 |
refseq_mrna_human_NM |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
29626 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P137045 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P33564466 |
- |
- |
Agilent Human 2 cDNA Microarray (G4101A) |
2054386 |
- |
- |
Agilent Human 2 cDNA Microarray (G4101A) |
2054386 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09388156 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09388138 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16716205 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09388151 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09388154 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09388133 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16716199 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16716201 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09388148 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09388160 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09388143 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09388159 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09388163 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09388146 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09388158 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16716208 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P09388145 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16716202 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P137045 |
- |
- |