Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002114.hg.1 |
ENST00000216962.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002114.hg.1 |
ENST00000471359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002114.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002114.hg.1 |
TC20000168.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000428458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000428458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000428458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000428458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000428458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000428458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000471359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000471359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000471359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000471359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000471359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
ENST00000471359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000147870.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147870.hg.1 |
ENST00000428458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147870.hg.1 |
ENST00000471359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147870.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000147870.hg.1 |
TC2000007015.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000147870.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147870.hg.1 |
ENST00000428458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147870.hg.1 |
ENST00000471359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147870.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000216962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000428458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000428458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000428458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000428458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000428458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000428458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000471359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000471359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000471359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000471359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000471359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
ENST00000471359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007015.hg.1 |
NM_002862.3 |
Refseq |