Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036782.hg.1 |
TC01002386.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036782.hg.1 |
ENST00000374272.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036782.hg.1 |
NM_032588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013374.hg.1 |
ENST00000374272.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013374.hg.1 |
ENST00000374272.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013374.hg.1 |
ENST00000374272.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013374.hg.1 |
ENST00000374272.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013374.hg.1 |
ENST00000374272.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013374.hg.1 |
ENST00000374272.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013374.hg.1 |
ENST00000374272.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013374.hg.1 |
ENST00000374272.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013374.hg.1 |
ENST00000374272.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013374.hg.1 |
ENST00000374272.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013374.hg.1 |
ENST00000483052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013374.hg.1 |
ENST00000483052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013374.hg.1 |
ENST00000483052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013374.hg.1 |
ENST00000483052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013374.hg.1 |
ENST00000483052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013374.hg.1 |
NM_032588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013374.hg.1 |
NM_032588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013374.hg.1 |
NM_032588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013374.hg.1 |
NM_032588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013374.hg.1 |
NM_032588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013374.hg.1 |
NM_032588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013374.hg.1 |
NM_032588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013374.hg.1 |
NM_032588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013374.hg.1 |
NM_032588.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013374.hg.1 |
NM_032588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100199357.hg.1 |
ENST00000374272.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100199357.hg.1 |
NM_032588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100199357.hg.1 |
ENST00000374272.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100199357.hg.1 |
NM_032588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100199357.hg.1 |
TC0100013374.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100013374.hg.1 |
ENST00000374272.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013374.hg.1 |
ENST00000374272.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013374.hg.1 |
ENST00000374272.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013374.hg.1 |
ENST00000374272.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013374.hg.1 |
ENST00000374272.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013374.hg.1 |
ENST00000374272.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013374.hg.1 |
ENST00000374272.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013374.hg.1 |
ENST00000374272.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013374.hg.1 |
ENST00000374272.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013374.hg.1 |
ENST00000374272.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013374.hg.1 |
ENST00000483052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013374.hg.1 |
ENST00000483052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013374.hg.1 |
ENST00000483052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013374.hg.1 |
ENST00000483052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013374.hg.1 |
ENST00000483052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013374.hg.1 |
NM_032588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013374.hg.1 |
NM_032588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013374.hg.1 |
NM_032588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013374.hg.1 |
NM_032588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013374.hg.1 |
NM_032588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013374.hg.1 |
NM_032588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013374.hg.1 |
NM_032588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013374.hg.1 |
NM_032588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013374.hg.1 |
NM_032588.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013374.hg.1 |
NM_032588.3 |
Refseq |